Learn More
Cayman Chemical Ganglioside GM1 (ovine) (ammonium salt), Purity: ≥98%, CAS: 1007119-81-4, 50 mg
SDP
Catalog No. 502991870
Encompass_Preferred
A ganglioside; enriched in lipid rafts; associates with growth factor receptors and is required for TrkA expression on the cell surface; interacts with other proteins to increase calcium influx, affecting various calcium-dependent processes, including inducing neuronal outgrowth during differentiation; acts as a receptor for cholera toxin and the heat-labile enterotoxin from E. coli; accumulates in neurons in ganglioside GM1 gangliosidosis; levels are decreased in the substantia nigra pars compacta in postmortem brain from patients with Parkinson’s disease
Catalog No. 50-299-1870 Supplier Cayman Chemical Supplier No. 3154950 MG
A ganglioside; enriched in lipid rafts; associates with growth factor receptors and is required for TrkA expression on the cell surface; interacts with other proteins to increase calcium influx, affecting various calcium-dependent processes, including inducing neuronal outgrowth during differentiation; acts as a receptor for cholera toxin and the heat-labile enterotoxin from E. coli; accumulates in neurons in ganglioside GM1 gangliosidosis; levels are decreased in the substantia nigra pars compacta in postmortem brain from patients with Parkinson’s disease