Learn More
Cayman Chemical GnglIosIde GM3bovIn braIn 1mg
SDP
Catalog No. 502969387
Encompass_Preferred
A ganglioside GM3 with C18:1 and C20:1 sphingoid backbones; demonstrates antiproliferative and pro-apoptotic effects in tumor cells by modulating cell adhesion, proliferation, and differentiation; suppresses angiogenesis and reduces proliferation and migration of HUVECs at 20 μM; induces dissociation of the insulin receptor-caveolin-1 complex from lipid microdomains
Catalog No. 50-296-9387 Supplier Cayman Chemical Supplier No. 155871 MG
A ganglioside GM3 with C18:1 and C20:1 sphingoid backbones; demonstrates antiproliferative and pro-apoptotic effects in tumor cells by modulating cell adhesion, proliferation, and differentiation; suppresses angiogenesis and reduces proliferation and migration of HUVECs at 20 μM; induces dissociation of the insulin receptor-caveolin-1 complex from lipid microdomains