Learn More
Cayman Chemical GnglIosIde GQ1bporcIna 100ug
SDP
Catalog No. 502990634
Encompass_Preferred
A sphingolipid molecular species isolated from porcine brain with primarily C18:0 fatty acyl chain lengths; stimulates phosphorylation of several ecto-type protein kinase substrates on the surface of GOTO human neuroblastoma cells at 5 nM; promotes differentiation of mESCs to neuronal precursor and glial cells via activation of the ERK1/2 pathway; induces differentiation of murine keratinocytes through phosphoinositide turnover
Catalog No. 50-299-0634 Supplier Cayman Chemical Supplier No. 31562100 UG
A sphingolipid molecular species isolated from porcine brain with primarily C18:0 fatty acyl chain lengths; stimulates phosphorylation of several ecto-type protein kinase substrates on the surface of GOTO human neuroblastoma cells at 5 nM; promotes differentiation of mESCs to neuronal precursor and glial cells via activation of the ERK1/2 pathway; induces differentiation of murine keratinocytes through phosphoinositide turnover