Learn More
Cayman Chemical GnglIosIde MIxture 1mL
SDP
Supplier:  Cayman Chemical 293621 ML
Encompass_Preferred
A mixture of gangliosides; contains the gangliosides asialo GM1, GM1, GD1a, GD1B, and GT1b; intended for use as a reference standard for the detection of these gangliosides in various samples by TLC, LC-, or GC-MS
Catalog No. 50-298-9195