Learn More
ABclonal Technology GNGT1 Rabbit pAb
SDP
Catalog No. 5000108117
Encompass_Preferred
This gene encodes the gamma subunit of transducin, a guanine nucleotide-binding protein (G protein) that is found in rod outer segments. Transducin, also known as GMPase, mediates the activation of a cyclic GTP-specific (guanosine monophosphate) phosphodiesterase by rhodopsin.
Catalog No. 50-001-08117 Supplier ABclonal Technology Supplier No. A15675500UL
This gene encodes the gamma subunit of transducin, a guanine nucleotide-binding protein (G protein) that is found in rod outer segments. Transducin, also known as GMPase, mediates the activation of a cyclic GTP-specific (guanosine monophosphate) phosphodiesterase by rhodopsin.