Learn More
Aviva Systems Biology Corp GNGT2 Antibody - middle region (ARP55401_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55401_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GNGT2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GNGT2. Alternate names: G-GAMMA-8, G-GAMMA-C, GNG8, GNG9, GNGT8ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-61491