Learn More
Cayman Chemical Bilobalide, Purity: ≥95%, CAS: 33570-04-6, 10 mg
SDP
Catalog No. 502954435
Encompass_Preferred
A sesquiterpene lactone which protects against cerebral edema, decreases cortical infarct volume, and reduces cerebral ischemic damage; at 10 μM, reduces the release of glycine and glutamate from hippocampal slices under ischemic conditions; increases the levels and activities of several cytochrome P450 isoforms in rat liver microsomes
Catalog No. 50-295-4435 Supplier Cayman Chemical Supplier No. 1427210 MG
A sesquiterpene lactone which protects against cerebral edema, decreases cortical infarct volume, and reduces cerebral ischemic damage; at 10 μM, reduces the release of glycine and glutamate from hippocampal slices under ischemic conditions; increases the levels and activities of several cytochrome P450 isoforms in rat liver microsomes