Learn More
Cayman Chemical Gnkgo bloba L Blobetn 10mg
SDP
Catalog No. 502941006
Encompass_Preferred
A biflavonoid with diverse biological activities; cytotoxic to HeLa, NCI-H460, Daudi, K562, SKOV3, MIA PaCa-2, and MCF-7 cells in vitro (IC50s = 14.79-97.28 μM); halts the cell cycle at the G2/M phase in HeLa cells in a concentration-dependent manner; induces apoptosis in HeLa cells at 20 μM; selectively inhibits MMP-9 (IC50 = 10.33 μM) over MMP-2 and MMP-3 (IC50s = >100 μM); inhibits aggregation of Aβ40 in vitro (IC50 = 4.7 μM)
Catalog No. 50-294-1006 Supplier Cayman Chemical Supplier No. 2789910 MG
A biflavonoid with diverse biological activities; cytotoxic to HeLa, NCI-H460, Daudi, K562, SKOV3, MIA PaCa-2, and MCF-7 cells in vitro (IC50s = 14.79-97.28 μM); halts the cell cycle at the G2/M phase in HeLa cells in a concentration-dependent manner; induces apoptosis in HeLa cells at 20 μM; selectively inhibits MMP-9 (IC50 = 10.33 μM) over MMP-2 and MMP-3 (IC50s = >100 μM); inhibits aggregation of Aβ40 in vitro (IC50 = 4.7 μM)