Learn More
Vector Laboratories Galanthus Nivalis Lectin (GNL), Fluorescein, 2mg
SDP
Supplier:  Vector Laboratories FL12412
Encompass
The recommended concentration range for use is 5-20 µg/ml. Solution: 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM CaCl2. Binding seems to be preferentially directed toward structures containing (α-1,3) mannose residues. Also in contrast to most mannose-binding lectins, GNL will not bind α-linked glucose. Reports indicate that this lectin binds rat and mouse IgM but not IgG. The only protein from human serum reported to bind to this lectin is α2-macroglobulin. GNL binds to many viral glycoproteins. Fluorescein labeled Galanthus nivalis lectin has an appropriate number of fluorochromes bound to provide the optimum staining characteristics for this lectin. This conjugate is supplied essentially free of unconjugated fluorochromes. The excitation maximum is at 495 nm and the emission maximum is at 515 nm.
Catalog No. 50-204-5658