Learn More
Aviva Systems Biology Corp GNL3L Antibody - N-terminal region (ARP58798_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58798_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GNL3L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GNL3L. Alternate names: FLJ10613, RP11-353K22.1ApplicationIHC, WB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62427