Learn More
Aviva Systems Biology Corp Gnmt Antibody - C-terminal region (ARP43565_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43565_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Gnmt. It was validated on Western Blot. . Alternate names: -ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-081