Learn More
Cayman Chemical GnoderenIc AcId D 10mg
SDP
Catalog No. 502944052
Encompass_Preferred
A triterpenoid with diverse biological activities; inhibits ACE (IC50 = 734 µM); cytotoxic to HepG2, HeLa, and Caco-2 cancer cells (IC50s = 0.14, 0.18, and 0.26 mg/ml, respectively); inhibits LPS-induced NO production in BV-2 microglia (IC50 = 13.77 µM)
Catalog No. 50-294-4052 Supplier Cayman Chemical Supplier No. 3977410 MG
A triterpenoid with diverse biological activities; inhibits ACE (IC50 = 734 µM); cytotoxic to HepG2, HeLa, and Caco-2 cancer cells (IC50s = 0.14, 0.18, and 0.26 mg/ml, respectively); inhibits LPS-induced NO production in BV-2 microglia (IC50 = 13.77 µM)