Learn More
Cayman Chemical Ganoderic Acid A, Purity: ≥95%, CAS: 81907-62-2, 50 mg
SDP
Catalog No. 502935281
Encompass_Preferred
A triterpene with diverse biological activities; reduces cell viability, expression SOD1, SOD2, and SOD3, and production of ROS in PC3 prostate cancer cells; inhibits LPS-induced activation of NF-κB, release of TNF-α, IL-1β, and IL-6, and mitochondrial activity in primary mouse cortical microglia; reduces lung MPO activity, neutrophil infiltration, and NF-κB signaling, as well as BALF levels of TNF-α, IL-1β, and IL-6 in a mouse model of LPS-induced lung injury at 20 and 40 mg/kg; reduces weight gain and hepatic lipid accumulation and improves insulin sensitivity in a mouse model of high-fat diet-induced obesity
Catalog No. 50-293-5281 Supplier Cayman Chemical Supplier No. 2667450 MG
A triterpene with diverse biological activities; reduces cell viability, expression SOD1, SOD2, and SOD3, and production of ROS in PC3 prostate cancer cells; inhibits LPS-induced activation of NF-κB, release of TNF-α, IL-1β, and IL-6, and mitochondrial activity in primary mouse cortical microglia; reduces lung MPO activity, neutrophil infiltration, and NF-κB signaling, as well as BALF levels of TNF-α, IL-1β, and IL-6 in a mouse model of LPS-induced lung injury at 20 and 40 mg/kg; reduces weight gain and hepatic lipid accumulation and improves insulin sensitivity in a mouse model of high-fat diet-induced obesity