Learn More
Cayman Chemical GnoderIc AcId G 10mg
SDP
Catalog No. 502946264
Encompass_Preferred
A triterpenoid with anticancer and anti-inflammatory activities; inhibits proliferation of Meth A sarcoma cells (IC50 = 6.8 UG/ml); inhibits the production of NO in LPS-induced BV-2 microglial cells (IC50 = 5.77 µM)
Catalog No. 50-294-6264 Supplier Cayman Chemical Supplier No. 3467510 MG
A triterpenoid with anticancer and anti-inflammatory activities; inhibits proliferation of Meth A sarcoma cells (IC50 = 6.8 UG/ml); inhibits the production of NO in LPS-induced BV-2 microglial cells (IC50 = 5.77 µM)