Learn More
Cayman Chemical GnoderIc AcId N 10mg
SDP
Catalog No. 502948380
Encompass_Preferred
A triterpene with enzyme inhibitory activity; inhibits ACE in a cell-free assay (IC50 = 0.941 mM)
Catalog No. 50-294-8380 Supplier Cayman Chemical Supplier No. 3432310 MG
A triterpene with enzyme inhibitory activity; inhibits ACE in a cell-free assay (IC50 = 0.941 mM)