Learn More
Cayman Chemical Ganoderol B, Purity: ≥95%, CAS: 104700-96-1, 10 mg
SDP
Catalog No. 502947440
Encompass_Preferred
A triterpenoid with diverse biological activities; inhibits ACE and α-glucosidase (IC50s = 220 and 119.8 µM, respectively); cytotoxic to HGC-27, HeLa, and A549 cells (IC50s = 16.15, 12.64, and 9.77 µM, respectively); prevents HSV-1 infection in Vero cells (EC50 = 68 µM); reduces testosterone-induced increases in ventral prostate weight in castrated rats at 0.001 and 0.1 mg/kg
Catalog No. 50-294-7440 Supplier Cayman Chemical Supplier No. 3432410 MG
A triterpenoid with diverse biological activities; inhibits ACE and α-glucosidase (IC50s = 220 and 119.8 µM, respectively); cytotoxic to HGC-27, HeLa, and A549 cells (IC50s = 16.15, 12.64, and 9.77 µM, respectively); prevents HSV-1 infection in Vero cells (EC50 = 68 µM); reduces testosterone-induced increases in ventral prostate weight in castrated rats at 0.001 and 0.1 mg/kg