Learn More
Innovative Research Inc GnPig RBCs 50ml
Catalog No. 502744415
Encompass_Preferred
Guinea Pig Red Blood Cells 50ml - Guinea Pig Red Blood Cells from Innovative Research are manufactured using whole blood collections from Guinea Pigs located in the USA. The whole blood is centrifuged to separate the platelet-rich plasma, buffy coat layer, and leukocytes. Red blood cells are perishable and are collected and processed upon receipt of order.
Catalog No. 50-274-4415 Supplier Innovative Research Inc Supplier No. IGPRBC50ML
Guinea Pig Red Blood Cells 50ml - Guinea Pig Red Blood Cells from Innovative Research are manufactured using whole blood collections from Guinea Pigs located in the USA. The whole blood is centrifuged to separate the platelet-rich plasma, buffy coat layer, and leukocytes. Red blood cells are perishable and are collected and processed upon receipt of order.