Learn More
Aviva Systems Biology Corp GNPTAB Antibody - N-terminal region (ARP49815_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49815_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GNPTAB. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human GNPTAB. Alternate names: DKFZp762B226, GNPTA, ICD, KIAA1208, MGC4170ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-454