Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 2798MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: F1G4; same as GNRH03. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: T47D cells. Pituitary gland, ovarian or breast cancers. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: A synthetic peptide aa 1-29 (MANSASPEQNQHCSAINNSIPLMQGNLPY) from the N-terminal of human GnRH receptor. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 54-60kDa.Chromosome: 4q21.2 (GNRHR) & 2p16.3 (LHCGR) (Human). Gene Symbol: GNRHR, LHCGR. Gene ID: 3973. Ensemble: ENSG00000138039. Uniprot: NP_000224.
Catalog No. 50-188-4748