Learn More
Electron Microscopy Sciences General Purpose Laboratory Microwave
SDP
Catalog No. 501931276
Encompass
Affordable general-purpose laboratory microwave oven features high-performance fume extraction and is ideal for general laboratory use including slide drying and histology staining. The Microwave features digital countdown timer accurate to 1 second, (5) built-in calibrated power settings, and stirrers (no carousel necessary) for even microwave distribution resulting in no "hot spots". The stainless steel interior and exterior provides for easy maintenance and long life. It is easy to use with "User friendly" controls and icon-based functions. Tall chambers allow for tall containers and vessels. Illuminated interiors with stain-resistant ceramic floors.
Catalog No. 50-193-1276 Supplier Electron Microscopy Sciences Supplier No. 97550
Affordable general-purpose laboratory microwave oven features high-performance fume extraction and is ideal for general laboratory use including slide drying and histology staining. The Microwave features digital countdown timer accurate to 1 second, (5) built-in calibrated power settings, and stirrers (no carousel necessary) for even microwave distribution resulting in no "hot spots". The stainless steel interior and exterior provides for easy maintenance and long life. It is easy to use with "User friendly" controls and icon-based functions. Tall chambers allow for tall containers and vessels. Illuminated interiors with stain-resistant ceramic floors.