Learn More
Aviva Systems Biology Corp GNS Antibody - C-terminal region (ARP45217_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45217_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GNS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human GNS. Alternate names: G6S, MGC21274ApplicationWB, IHC. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58984