Learn More
Aviva Systems Biology Corp Gns Antibody - C-terminal region (ARP45216_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45216_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Gns. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: GnsApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-190