Learn More
Cayman Chemical GNTIhydrochlorIde 10mg
SDP
Catalog No. 502988074
Encompass_Preferred
A κ-opioid receptor antagonist (Ki = 0.18 nM); selective for κ-opioid receptors over µ- and δ-opioid receptors in radioligand binding assays (Kis = 36.9 and 70 nM, respectively); inhibits agonist-induced [35S]GTPγS binding in CHO cells expressing human κ-opioid receptors (pA2 = 10.4); induces scratching behavior in mice at 0.3 mg/kg
Catalog No. 50-298-8074 Supplier Cayman Chemical Supplier No. 3159710 MG
A κ-opioid receptor antagonist (Ki = 0.18 nM); selective for κ-opioid receptors over µ- and δ-opioid receptors in radioligand binding assays (Kis = 36.9 and 70 nM, respectively); inhibits agonist-induced [35S]GTPγS binding in CHO cells expressing human κ-opioid receptors (pA2 = 10.4); induces scratching behavior in mice at 0.3 mg/kg