Learn More
Selleck Chemical LLC GO-203 2mg 1222186-26-6
SDP
Supplier:  Selleck Chemical LLC S86742MG
Encompass_Preferred
GO-203 is a D-amino acid cell-penetrating peptide inhibitor of MUC1-C dimerization and thereby its oncogenic function. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1534230