Learn More
Medchemexpress LLC GO-203 TFA | 1222186-26-6 | 98.39% | 2426.77 | 5 MG
SDP
Catalog No. 5000422359
Encompass_Preferred
GO-203 TFA is a potent inhibitor of the MUC1-C oncoprotein. It is an all D-amino acid peptide that contains a poly-R transduction domain linked to a CQCRRKN motif, which binds to the MUC1-C cytoplasmic tail and prevents MUC1-C homodimerization. This compound downregulates TIGAR protein synthesis by inhibiting the PI3K-AKT-S6K1 pathway. It also induces the production of Reactive Oxygen Species (ROS) and leads to a loss of mitochondrial transmembrane potential. GO-203 TFA has been shown to inhibit the growth of colon cancer cells both in vitro and as xenografts in nude mice.
  • Potent MUC1-C oncoprotein inhibitor
  • All D-amino acid peptide
  • Blocks MUC1-C homodimerization
  • Downregulates TIGAR protein synthesis by inhibiting the PI3K-AKT-S6K1 pathway
  • Induces ROS production and loss of mitochondrial transmembrane potential
  • Inhibits the growth of colon cancer cells in vitro and in xenografts in nude mice
Catalog No. 50-004-22359 Supplier Medchemexpress LLC Supplier No. HYP1925A5MG
GO-203 TFA is a potent inhibitor of the MUC1-C oncoprotein. It is an all D-amino acid peptide that contains a poly-R transduction domain linked to a CQCRRKN motif, which binds to the MUC1-C cytoplasmic tail and prevents MUC1-C homodimerization. This compound downregulates TIGAR protein synthesis by inhibiting the PI3K-AKT-S6K1 pathway. It also induces the production of Reactive Oxygen Species (ROS) and leads to a loss of mitochondrial transmembrane potential. GO-203 TFA has been shown to inhibit the growth of colon cancer cells both in vitro and as xenografts in nude mice.
  • Potent MUC1-C oncoprotein inhibitor
  • All D-amino acid peptide
  • Blocks MUC1-C homodimerization
  • Downregulates TIGAR protein synthesis by inhibiting the PI3K-AKT-S6K1 pathway
  • Induces ROS production and loss of mitochondrial transmembrane potential
  • Inhibits the growth of colon cancer cells in vitro and in xenografts in nude mice