Learn More
Apexbio Technology LLC Go 6976, 25mg Cas# 136194-77-9
SDP
Catalog No. 501998592
Encompass_Preferred
Go 6976 5mg. Go 6976 is a selective inhibitor of PKC with IC50 value of 20 nM [1] [2]. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8592 Supplier Apexbio Technology LLC Supplier No. A834125
Go 6976 5mg. Go 6976 is a selective inhibitor of PKC with IC50 value of 20 nM [1] [2]. There are other sizes available. Please inqury us for quote.