Learn More
Enzo Life Sciences Go-6983 (0.5mg). CAS: 133053-19-7
SDP
Catalog No. 502010679
Encompass_Preferred
PKC inhibitor. Has been shown to selectively inhibit several PKC isoenzymes (IC50 = 7nM for PKCα and PKCβ; 6nM for PKCγ; 10nM for PKCδ; 60nM for PKCζ). The compound does not effectively inhibit PKCµ (IC50=20 µM) and therefore can be used to differentiate PKCµ from other isoforms. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-0679 Supplier Enzo Life Sciences Supplier No. BMLEI2960500
PKC inhibitor. Has been shown to selectively inhibit several PKC isoenzymes (IC50 = 7nM for PKCα and PKCβ; 6nM for PKCγ; 10nM for PKCδ; 60nM for PKCζ). The compound does not effectively inhibit PKCµ (IC50=20 µM) and therefore can be used to differentiate PKCµ from other isoforms. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.