Learn More
Apexbio Technology LLC Go 6983(Synonyms: GO-6983, GO6983, PKC Inhibitor GO 6983, Pan-PKC Inhibitor GO-6983, PKC inhibitor GO6983, 133053-19-7), 10mM (in 1mL DMSO), CAS: 133053-19-7.
SDP
Catalog No. 5000060474
Encompass_Preferred
Go 6983 (CAS 133053-19-7) is a selective inhibitor of protein kinase C isoforms PKC PKC PKC PKC and PKC with reported IC50 values of approximately 7 nM 7 nM 6 nM 10 nM and 20 mM respectively Protein kinase C (PKC) isoforms act as receptors for tumor-promoting phorbol ester mediating various downstream cellular functions In cancer cell models Go 6983 suppresses PKC and PKC activation induced by phorbol esters and reduces PKC expression inhibiting cell survival pathways It is frequently employed in biomedical studies examining PKC-dependent signaling in cancer progression and epithelial-to-mesenchymal transition (EMT)
Catalog No. 50-000-60474 Supplier Apexbio Technology LLC Supplier No. A83435.1
Go 6983 (CAS 133053-19-7) is a selective inhibitor of protein kinase C isoforms PKC PKC PKC PKC and PKC with reported IC50 values of approximately 7 nM 7 nM 6 nM 10 nM and 20 mM respectively Protein kinase C (PKC) isoforms act as receptors for tumor-promoting phorbol ester mediating various downstream cellular functions In cancer cell models Go 6983 suppresses PKC and PKC activation induced by phorbol esters and reduces PKC expression inhibiting cell survival pathways It is frequently employed in biomedical studies examining PKC-dependent signaling in cancer progression and epithelial-to-mesenchymal transition (EMT)