Learn More
Vernier™ Go to Easy Adapter
SDPSCI_ED
Catalog No. S15996ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
Go!Temp and Go!Link
Catalog No. For Use With (Equipment)
S15996ND Go!Temp and Go!Link
1 options
Catalog No. S15996ND Supplier Vernier™ Supplier No. USBMINI
Add to Cart
Add to Cart
Includes
Go to Easy Adapter
This adapter allows a Go!Temp or Go!Link* to connect to the USB port of a TI-Nspire handheld or TI-84 graphing calculator. The adapter has a USB standard-A receptacle which connects to the Go!Temp or Go!Link and a USB Mini-A USB plug that connects to the calculator. The adapter can also be used with the USB cable that comes with a Go!Motion.
  • Connector 1: USB Standard Type A Receptacle
  • Connector 2: USB Mini Type A Plug
  • The adapter has a USB standard-A receptacle which connects to the Go!Temp or Go!Link and a USB Mini-A USB plug that connects to the calculator.
  • The adapter can also be used with the USB cable that comes with a Go!Motion.
Warranty and Services
5-year limited warranty
Specifications
For Use With (Equipment) Go!Temp and Go!Link
For Use With (Application) Educational use only
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov