Learn More
Immunoreagents Inc GOAT A-CAT IGG (H&L) AP 1 MG
Supplier:  Immunoreagents Inc GTXCT003D
Encompass_Preferred
Secondary Antibody; Goat anti-cat (Feline) IgG (H&L), Affinity Pure, Host: Goat, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC
Catalog No. 50-272-109