Learn More
Immunoreagents Inc GOAT A-CHICKEN IGG ALP 1 MG
Supplier:  Immunoreagents Inc GTXCK003DALP
Encompass_Preferred
Secondary Antibody; Goat anti-chicken IgG/Y (H&L), Affinity Pure, ALP Conjugate, Host: Goat, Format: Liq, Label: ALP, Applications: WB, ELISA
Catalog No. 50-271-968