Learn More
Immunoreagents Inc GOAT A-CHICKEN IGG AP 2 MG
Supplier:  Immunoreagents Inc GTXCK003D
Encompass_Preferred
Secondary Antibody; Goat anti-chicken IgG/Y (H&L), Affinity Pure, Host: Goat, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC
Catalog No. 50-270-763