Learn More
Immunoreagents Inc GOAT A-CHICKEN IGG HRP 1 MG
Supplier:  Immunoreagents Inc GTXCK003DHRPX
Encompass_Preferred
Secondary Antibody; Goat anti-chicken IgG/Y (H&L), Affinity Pure, HRP Conjugate, Host: Goat, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-270-787