Learn More
Immunoreagents Inc GOAT A-DOG IGG BIO 0.5 MG
Supplier:  Immunoreagents Inc GTXDG003DBIO
Encompass_Preferred
Secondary Antibody; Goat anti-dog (Canine) IgG (H&L), Affinity Pure, Biotin Conjugate, Host: Goat, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-302