Learn More
Immunoreagents Inc GOAT A-GP IGG DYLIGHT 650 1MG
Supplier:  Immunoreagents Inc GTXGP003D650NHSX
Encompass_Preferred
Secondary Antibody; Goat anti-guinea pig IgG (H&L), Affinity Pure, DyLight 650 Conjugate, Host: Goat, Format: Lyo, Label: DyL650, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-270-920