Learn More
Immunoreagents Inc GOAT A-GUINEAPIG IGG FITC 1MG
Supplier:  Immunoreagents Inc GTXGP003FFITC
Encompass_Preferred
Secondary Antibody; Goat anti-guinea pig IgG (H&L), Affinity Pure, min x w/Bo, Ck, Gt, Ha, EQ, Hu, Mu, Rb, Rt or Sh serum proteins, FITC Conjugate, Host: Goat, Format: Liq, Label: FITC, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-270-922