Learn More
Immunoreagents Inc GOAT A-HUMAN IG KAPPA BIO
Supplier:  Immunoreagents Inc GTXHU084EBIO
Encompass_Preferred
Secondary Antibody; Goat anti-human (kappa light chain) - Affinity Pure, min x w/Mu Serum Proteins, Biotin Conjugate, Host: Goat, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-720