Learn More
Immunoreagents Inc GOAT A-HUMAN IG KAPPA DYLIT650
Supplier:  Immunoreagents Inc GTXHU084E650NHSX
Encompass_Preferred
Secondary Antibody; Goat anti-human (kappa light chain) - Affinity Pure, min x w/Mu Serum Proteins, DyLight 650 Conjugate, Host: Goat, Format: Lyo, Label: DyL650, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-271-725