Learn More
Immunoreagents Inc GOAT A-HUMAN IG KAPPA HRP
Supplier:  Immunoreagents Inc GTXHU084EHRPX
Encompass_Preferred
Secondary Antibody; Goat anti-human (kappa light chain) - Affinity Pure, min x w/Mu Serum Proteins, HRP Conjugate, Host: Goat, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-266