Learn More
Immunoreagents Inc GOAT A-HUMAN IG KAPPA HRP
Supplier:  Immunoreagents Inc GTXHU084DHRPX
Encompass_Preferred
Secondary Antibody; Goat anti-human (kappa light chain) - Affinity Pure, HRP Conjugate, Host: Goat, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-256