Learn More
Immunoreagents Inc GOAT A-HUMAN IGA AP 1 MG
Catalog No. 50270767
Encompass_Preferred
Secondary Antibody; Goat anti-human IgA Alpha Chain, Affinity Pure, Host: Goat, Format: Liq, Label: None , Applications: WB, ELISA, IHC/ICC, IF, IP, FC
Catalog No. 50-270-767 Supplier Immunoreagents Inc Supplier No. GTXHU001E
Secondary Antibody; Goat anti-human IgA Alpha Chain, Affinity Pure, Host: Goat, Format: Liq, Label: None , Applications: WB, ELISA, IHC/ICC, IF, IP, FC