Learn More
Immunoreagents Inc GOAT A-HUMAN IGA BIO 0.5 MG
Supplier:  Immunoreagents Inc GTXHU001GBIO
Encompass_Preferred
Secondary Antibody; Goat anti-human IgA Alpha Chain, F(ab)'2 fragment - Affinity Pure, Biotin Conjugate, Host: Goat, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-996