Learn More
Immunoreagents Inc GOAT A-HUMAN IGA DYLIGHT 800
Supplier:  Immunoreagents Inc GTXHU001D800NHSX
Encompass_Preferred
Secondary Antibody; Goat anti-human IgA Alpha Chain, Affinity Pure, DyLight 800 Conjugate, Host: Goat, Format: Lyo, Label: DyL800, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-271-450