Learn More
Immunoreagents Inc GOAT A-HUMAN IGA HRP 0.5 MG
Supplier:  Immunoreagents Inc GTXHU001GHRPX
Encompass_Preferred
Secondary Antibody; Goat anti-human IgA Alpha Chain, F(ab)'2 fragment - Affinity Pure, HRP Conjugate, Host: Goat, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-918