Learn More
Immunoreagents Inc GOAT A-HUMAN IGG/A/M ALP
Supplier:  Immunoreagents Inc GTXHU010GALP
Encompass_Preferred
Secondary Antibody; Goat anti-human IgG + IgA + IgM, F(ab)'2 fragment - Affinity Pure, ALP Conjugate, Host: Goat, Format: Liq, Label: ALP, Applications: WB, ELISA
Catalog No. 50-271-513