Learn More
Immunoreagents Inc GOAT A-HUMAN IGG/A/M AP
Catalog No. 50270777
Encompass_Preferred
Secondary Antibody; Goat anti-human IgG + IgA + IgM, F(ab)'2 fragment - Affinity Pure, Host: Goat, Format: Liq, Label: None , Applications: WB, ELISA, IHC/ICC, IF, IP, FC
Catalog No. 50-270-777 Supplier Immunoreagents Inc Supplier No. GTXHU010G
Secondary Antibody; Goat anti-human IgG + IgA + IgM, F(ab)'2 fragment - Affinity Pure, Host: Goat, Format: Liq, Label: None , Applications: WB, ELISA, IHC/ICC, IF, IP, FC