Learn More
Immunoreagents Inc GOAT A-HUMAN IGG/A/M BIO
Supplier:  Immunoreagents Inc GTXHU010GBIO
Encompass_Preferred
Secondary Antibody; Goat anti-human IgG + IgA + IgM, F(ab)'2 fragment - Affinity Pure, Biotin Conjugate, Host: Goat, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-514