Learn More
Immunoreagents Inc GOAT A-HUMAN IGG/A/M FITC
Supplier:  Immunoreagents Inc GTXHU010GFITC
Encompass_Preferred
Secondary Antibody; Goat anti-human IgG + IgA + IgM, F(ab)'2 fragment - Affinity Pure, FITC Conjugate, Host: Goat, Format: Liq, Label: FITC, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-271-137