Learn More
Immunoreagents Inc GOAT A-HUMAN IGG/A/M HRP
Supplier:  Immunoreagents Inc GTXHU010DHRPX
Encompass_Preferred
Secondary Antibody; Goat anti-human IgG + IgA + IgM - Affinity Pure, HRP Conjugate, Host: Goat, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-270-698