Learn More
Immunoreagents Inc GOAT A-HUMAN IGG AP 1 MG
Catalog No. 50270792
Encompass_Preferred
Secondary Antibody; Goat anti-human IgG, Fc - Affinity Pure, min x w/Bo, Eq, Mu or Rb serum proteins or Mu IgG1, Host: Goat, Format: Liq, Label: None , Applications: WB, ELISA, IHC/ICC, IF, IP, FC
Catalog No. 50-270-792 Supplier Immunoreagents Inc Supplier No. GTXHU004E2
Secondary Antibody; Goat anti-human IgG, Fc - Affinity Pure, min x w/Bo, Eq, Mu or Rb serum proteins or Mu IgG1, Host: Goat, Format: Liq, Label: None , Applications: WB, ELISA, IHC/ICC, IF, IP, FC